Läs mer
DMP 696, MedChemExpress
Produktkod. 16447727
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16447727 5 mg
16414998 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16447727 Leverantör Medchem Express Leverantörsnummer HY12131
MedChemExpress DMP 696 is a selective corticotropin-releasing hormone receptor 1 (CRHR1) antagonist, used for the treatment of anxiety and depression.
DMP 696 is a selective corticotropin-releasing hormone receptor 1 (CRHR1) antagonist, used for the treatment of anxiety and depression.
Specifikationer
Kemiskt namn eller material DMP 696
Fysisk form Powder
CAS 202578-52-7
Färg Light Brown
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C18H21Cl2N5O2
För användning med (applikation) Metabolism-protein/nucleotide metabolism
Löslighetsinformation DMSO : 100 mg/mL (243.72 mM; Need ultrasonic)
LEDER CC1=NN2C(N=C(C)N=C2NC(COC)COC)=C1C3=CC=C(Cl)C=C3Cl
Molekylvikt (g/mol) 410.3
Formel vikt 410.3
Procent renhet 98.73%