missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DMWD (Human) Recombinant Protein
Human DMWD full-length ORF ( AAH19266.1, 1 a.a. - 334 a.a.) recombinant protein with GST-tag at N-terminal.
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH19266.1 |
---|---|
Gene ID (Entrez) | 1762 |
Name | dystrophia myotonica, WD repeat containing |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16107284
|
Abnova™
H00001762-P01.10UG |
10 μg |
4515.00 SEK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16117284
|
Abnova™
H00001762-P01.25UG |
25 μg |
6845.00 SEK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Sequence: MLLRGLMKSYFGGLLCVCWSPDGRYVVTGGEDDLVTVWSFTEGRVVARGHGHKSWVNAVAFDPYTTRAEEAATAAGADGERSGEEEEEEPEAAGTGSAGGAPLSPLPKAGSITYRFGSAGQDTQFCLWDLTEDVLYPHPPLARTRTLPGTPGTTPPAASSSRGGEPGPGPLPRSLSRSNSLPHPAGGGKAGGPGVAAEPGTPFSIGRFATLTLQERRDRGAEKEHKRYHSLGNISRGGSGGSGSGGEKPSGPVPRSRLDPAKVLGTALCPRIHEVPLLEPLVCKKIAQERLTVLLFLEDCIITACQEGLICTWARPGKAGISSQPGNSPSGTVV
Specifications
AAH19266.1 | |
dystrophia myotonica, WD repeat containing | |
125% SDS-PAGE Stained with Coomassie Blue | |
D19S593E, DMR-N9, DMRN9, gene59 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
1762 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DMWD | |
GST |