missing translation for 'onlineSavingsMsg'
Learn More

DNA, Fish Sperm, Sodium Salt, Thermo Scientific™

Catalog Number p-7082747
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
9007-49-2
C17H15BrFNO3
50000-100000
MFCD00130922
GXZCYECDYOBPGH-UHFFFAOYNA-N
73904662
*
This item is not returnable. View return policy
9007-49-2
C17H15BrFNO3
50000-100000
MFCD00130922
GXZCYECDYOBPGH-UHFFFAOYNA-N
73904662
*
Physical Form Powder
Quantity 100 g
IUPAC Name (2 R)-3-(3-bromophenyl)-2-[[2-(4-fluorophenyl)acetyl]amino]propanoic acid
Packaging Plastic bottle
Chemical Name or Material DNA

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'