missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
DNA-PK Monoclonal Antibody (42-psc (42-26, 42-27)), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513249
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
MA5-13249 targets DNA-PKcs in GS, IF, and IHC (P) applications and shows reactivity with Human samples. The MA5-13249 immunogen is human DNA-dependent Protein Kinase purified from HeLa cells.
DNA-PK is a heterotrimeric, DNA-dependent protein kinase that initiates signaling cascades in response to DNA damage. DNA-PK is an atypical kinase, closely related to the PI3 kinases, and is often potently inhibited by small molecules designed to target PI3 kinases. DNA-PK encodes a component of the mediator complex, a transcriptional coactivator complex thought to be required for the expression of almost all genes. The mediator complex is recruited by transcriptional activators or nuclear receptors to induce gene expression, possibly by interacting with RNA polymerase II and promoting the formation of a transcriptional pre-initiation complex. Multiple transcript variants encoding different isoforms have been found for DNA-PK.
Specifikationer
| DNA-PK | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P78527 | |
| PRKDC | |
| Human DNA-dependent Protein Kinase purified from HeLa cells | |
| 500 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Gel Shift, Immunohistochemistry (Paraffin) | |
| 42-psc (42-26, 42-27) | |
| Unconjugated | |
| PRKDC | |
| AI326420; AU019811; DNA-dependent protein kinase catalytic subunit; DNAPDcs; DNAPK; DNA-PK catalytic subunit; DNA-PKcs; DNPK1; DOXNPH; doxorubicin nephropathy; dxnph; hyper-radiosensitivity of murine scid mutation, complementing 1; HYRC; HYRC1; I79_017613; IMD26; p350; p460; Prkdc; protein kinase, DNA activated, catalytic polypeptide; protein kinase, DNA-activated, catalytic polypeptide; scid; severe combined immunodeficiency; slip; Xrcc7 | |
| Mouse | |
| Protein G | |
| RUO | |
| 5591 | |
| 4° C | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering