missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAH14 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25072-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Force generating protein of respiratory cilia. Produces force towards the minus ends of microtubules. Dynein has ATPase activity; the force-producing power stroke is thought to occur on release of ADP. Involved in sperm motility; implicated in sperm flagellar assembly.Specifications
DNAH14 | |
Polyclonal | |
Unconjugated | |
DNAH14 | |
C1orf67, Ciliary dynein heavy chain 14, DNAH14, Dnahc14, HL 18, HL18 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
127602 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q0VDD8 | |
DNAH14 | |
DNAH14 Fusion Protein Ag18669 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |