Läs mer
Invitrogen™ DNAI1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588366
Beskrivning
Immunogen sequence: VGYGSYDFMK QSRGMLLLYS LKNPSFPEYM FSSNSGVMCL DIHVDHPYLV AVGHYDGNVA IYNLKKPHSQ PSFCSSAKSG KHSDPVWQVK WQKDDMDQNL NFFSVSSDGR IVSWTLVKRK LVHIDVIKLK VEGSTTEVPE GLQLHPVGCG TAFDFHKEID YMFLVGTEEG KIYKCSKSYS SQFLDTYDAH NMSVDTVSWN PYHTKVFMSC SSDWTVKIWD HTIKTPMFIY DLNSAVGDVA WAPYSSTVFA AVTTDGKAHI FDLAINKYEA ICNQPVAAKK NRLTHVQFNL IHPIIIVGDD RGHIISLKLS PNLRKMPKEK KGQEVQKGPA VEIAKLDKLL NLVREVKIKT; Positive Samples: HeLa, SKOV3, U-87MG, Mouse kidney; Cellular Location: Cytoplasm, cilium axoneme, cytoskeleton.
Eukaryotic cells rely on actin and microtubule-based protein motors to generate intracellular movements. These protein motors contain specialized domains that hydrolyze ATP to produce force and movement along a cytoskeletal polymer (actin in the case of myosin family and microtubules in the case of the kinesin family and dyneins). Dynein has been implicated in cytoplasmic motile functions, including chromosomal movement, retrograde organelle and axonal transport, the endocytic pathway, and the organization of the Golgi apparatus. In all cell types, dynein has the same basic structures and is composed of two or three distinct heavy chains (~450 kDa), three intermediate chains (70-125 kDa), and at least four light chains (15-25 kDa).
Specifikationer
| DNAI1 | |
| Polyclonal | |
| Unconjugated | |
| DNAI1 | |
| 1110066F04Rik; axonemal dynein intermediate chain 1; b2b1526Clo; BB124644; CILD1; DIC1; Dnai1; Dnaic1; dynein axonemal intermediate chain 1; Dynein intermediate chain 1, axonemal; dynein, axonemal, intermediate chain 1; dynein, axonemal, intermediate polypeptide 1; I79_025143; ICS; ICS1; immotile cilia syndrome 1; MGC26204; PCD; RP11-296L22.2; testis tissue sperm-binding protein Li 87P | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27019 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.56 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9UI46 | |
| DNAI1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 350-699 of human DNAI1 (NP_0362761). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.