missing translation for 'onlineSavingsMsg'
Learn More

DNAJA1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11713-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16868973

  • 4721.67 SEK / 150µL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa.
Specifications

Specifications

DNAJA1
Polyclonal
Unconjugated
DNAJA1
DJ 2, DjA1, DnaJ protein homolog 2, DNAJ2, DNAJA1, hDJ 2, HDJ2, Heat shock 40 kDa protein 4, Heat shock protein J2, HSDJ, HSJ 2, HSJ2, HSPF4, Human DnaJ protein 2
Rabbit
Antigen Affinity Chromatography
RUO
15502, 3301
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P31689, P63037
Dnaja1
DNAJA1 Fusion Protein Ag2284
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers