Learn More
DNAJB1 Mouse anti-Human, Unconjugated, Clone: 1E8E10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67422-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16840046
  • 1236.20 SEK / 20µL
Estimated Shipment: 03-10-2022
to see stock.
Description
Description
The DNAJB1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DNAJB1. This antibody recognizes human antigen. The DNAJB1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
DNAJB1
Monoclonal
500 μg/mL
Mouse
Primary
Human
ELISA, Western Blot
1E8E10
Unconjugated
20 μL
3337
IgG1