missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJB8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17071-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
DNAJB8 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain.Specifications
DNAJB8 | |
Polyclonal | |
Unconjugated | |
DNAJB8 | |
DJ6, DNAJB8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
165721, 500253, 56691 | |
-20°C | |
Liquid |
Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8NHS0, Q9QYI7 | |
DNAJB8 | |
DNAJB8 Fusion Protein Ag10899 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |