Läs mer
R&D Systems™ DNAJC19 Overexpression Lysate
Produktkod. 18231401 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18231401 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Produktkod. 18231401 Leverantör R&D Systems™ Leverantörsnummer NBL109950
 Begränsad artikel
Beräknad leverans: 01-12-2026
Lägg i varukorgen
Lägg i varukorgen
missing translation for 'validMainframeMsg'
A Novus product, part of R&D Systems.
The DNAJC19 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Specifica
Celllinje The lysate was created in HEK293T cells, using plasmid ID RC2991 and based on accession number NM_145261. The protein contains a C-terminal DDK tag.
Kvantitet 0.1 mg
Förvaringsbuffert RIPA buffer
Renhet Protein
Testspecificitet Homo sapiens DnaJ (Hsp40) homolog, subfamily C, member 19 (DNAJC19), mRNA.
Innehåll och lagring -80°C. Avoid freeze-thaw cycles.
Produktlinje Novus
Proteintillstånd Native
För användning med (applikation) Western Blot
For Research Use Only
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.