missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | DNAL1 |
---|---|
Concentration | 0.15 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16835164
|
Proteintech
15809-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 18-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16825164
|
Proteintech
15809-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 18-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
DNAL1 was identified as a potential candidate gene for primary ciliary dyskinesia (PCD), a genetically heterologous disorder characterized by chronic infections of the upper and lower airways that often leads to permanent lung damage, randomization of left/right body symmetry, and reduced fertility. DNAL1 is reported to be expressed solely in tissues carrying motile cilia for flagella and interacts with DNAH5, a protein that when mutated has been shown to result in PCD. It has been suggested that DNAL1 serves a regulatory function for DNAH5 activity in outer dynein arms of sperm flagella, respiratory cilia, and ependymal cilia. DNAL1 has also been recently identified as an HIV dependency factor (HDF), suggesting that DNAL1 may be an important drug target in HIV treatment. At least two isoforms of DNAL1 are known to exist.Specifications
DNAL1 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q05A62, Q4LDG9 | |
105000, 685664, 83544 | |
DNAL1 Fusion Protein Ag8545 | |
Primary | |
-20°C | |
DNAL1 |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C14orf168, DNAL1, Dynein light chain 1, axonemal | |
Dnal1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |