missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNALI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17601-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene is the human homolog of the Chlamydomonas inner dynein arm gene, p28. The precise function of this gene is not known, however, it is a potential candidate for immotile cilia syndrome (ICS). Ultrastructural defects of the inner dynein arms are seen in patients with ICS. Immotile mutant strains of Chlamydomonas, a biflagellated algae, exhibit similar defects.Specifications
DNALI1 | |
Polyclonal | |
Unconjugated | |
DNALI1 | |
DNALI1, hp28, P28 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
298524, 75563, 7802 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O14645, Q4FZV3, Q8BVN8 | |
Dnali1 | |
DNALI1 Fusion Protein Ag11652 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |