missing translation for 'onlineSavingsMsg'
Learn More

DNMT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 19221-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16878924

  • 1656.73 SEK / 20µL
Estimated Shipment: 17-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

DNMT2 encodes a member of the solute carrier family 11 protein family. The product of this gene transports divalent metals and is involved in iron absorption. Mutations in this gene are associated with hypochromic microcytic anemia with iron overload. A related solute carrier family 11 protein gene is located on chromosome 2. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

DNMT2
Polyclonal
Unconjugated
TRDMT1
DNA MTase homolog HsaIIP, DNMT2, M.HsaIIP, PuMet, RNMT1, TRDMT1
Rabbit
Antigen Affinity Chromatography
RUO
13434, 1787, 291324
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O14717, O55055, Q4G073
Trdmt1
DNMT2 Fusion Protein Ag5571
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers