Läs mer
DO34, MedChemExpress
Produktkod. 16497637
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
Förpackningsstorlek:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16497637 5 mg 5mg
16464888 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16497637 Leverantör Medchem Express Leverantörsnummer HY1177715mg
MedChemExpress DO34 is a highly potent, selective and centrally active diacylglycerol lipase (DAGL) inhibitor, with an IC50 of 6 nM for DAGLα conversion of SAG to 2-AG, and an IC50 for DAGLβ.
DO34 is a highly potent, selective and centrally active diacylglycerol lipase (DAGL) inhibitor, with an IC50 of 6 nM for DAGLα conversion of SAG to 2-AG, and an IC50 for DAGLβ.
Specifikationer
Kemiskt namn eller material DO34
CAS 1848233-58-8
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C26H28F3N5O4
Kvantitet 5 mg
Löslighetsinformation DMSO : ≥ 100 mg/mL (188.14 mM)
LEDER O=C(N1CC(CC2=CC=CC=C2)N(C(N3N=NC(C4=CC=C(OC(F)(F)F)C=C4)=C3)=O)CC1)OC(C)(C)C
Molekylvikt (g/mol) 531.53
Formel vikt 531.53
Procent renhet 98.18%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Metabolism-protein/nucleotide metabolism