missing translation for 'onlineSavingsMsg'
Läs mer

DO3A, TRC

Produktkod. 30133667
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133667 50 mg
30137128 100 mg
30009820 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30133667 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD49488550MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 217973-03-0
Molekylformel C14H23N4O6 • 3(Na)
Molekylvikt (g/mol) 412.32
Synonym Trisodium 1,4,7,10-Tetraazacyclododecane-1,4,7-triacetate, 1,4,7,10-Tetraazacyclododecane-1,4,7-triacetic Acid Trisodium Salt
IUPAC-namn sodium 2,2',2' '-(1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetate
LEDER O=C(O[Na])CN1CCN(CC(O[Na])=O)CCN(CC(O[Na])=O)CCNCC1

Specifikationer

Kemiskt namn eller material DO3A
Kvantitet 50 mg
InChI formel InChI=1S/C14H26N4O6.3Na/c19-12(20)9-16-3-1-15-2-4-17(10-13(21)22)6-8-18(7-5-16)11-14(23)24;;;/h15H,1-11H2,(H,19,20)(H,21,22)(H,23,24);;;/q;3*+1/p-3
Formel vikt 412.13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.