missing translation for 'onlineSavingsMsg'
Learn More

DOC2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20574-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16840135

  • 3313.85 SEK / 150µL
Estimated Shipment: 03-10-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The antibody is specific to DOC2B.

There are at least two protein isoforms of the Double C2 protein, namely alpha and beta , which contain two C2-like domains. DOC2A and DOC2B are encoded by different genes; these genes are at times confused with the unrelated DAB2 gene which was initially named DOC-2. DOC2B is expressed ubiquitously and is suggested to be involved in Ca-dependent intracellular vesicle trafficking in various types of cells.
Specifications

Specifications

DOC2B
Polyclonal
Unconjugated
DOC2B
Doc2 beta, DOC2B, DOC2BL, double C2 like domains, beta
Rabbit
Antigen Affinity Chromatography
RUO
13447, 81820, 8447
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P70169, P70610, Q14184
Doc2b
Peptide
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers