Läs mer
Docetaxel Trihydrate, MedChemExpress
Beskrivning
Docetaxel Trihydrate (RP-56976 Trihydrate) is an antineoplastic agent and inhibits microtubule depolymerization with an IC50 value of 0.2 μM. Docetaxel Trihydrate is a semisynthetic analog of taxol and attenuates the effects of bcl-2 and bcl-xL gene expression. Docetaxel Trihydrate arrests the cell cycle at G2/M and leads to cell apoptosis.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Docetaxel Trihydrate |
| CAS | 148408-66-6 |
| Färg | White |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C43H59NO17 |
| Kvantitet | 100 mg |
| Synonym | RP-56976 Trihydrate |
| Löslighetsinformation | DMSO : 250 mg/mL (290.05 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (58.01 mM; Need ultrasonic) |
| LEDER | O=C(OC(C)(C)C)N[C@H]([C@H](C(O[C@@H]1C(C)=C([C@@H](O)C([C@@]2(C)[C@]([C@@](CO3)(OC(C)=O)[C@@]3([H])C[C@@H]2O)([H])[C@@H]4OC(C5=CC=CC=C5)=O)=O)C(C)(C)[C@@]4(O)C1)=O)O)C6=CC=CC=C6.O.O.O |
| Molekylvikt (g/mol) | 861.93 |
| Visa mer |