missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOCK10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16834-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. Overexpression of this gene results in a decrease in the processing of insulin and hepatocyte growth factor receptors to their mature subunits. This decrease is caused by the mislocalization of furin, the endoprotease responsible for cleavage of insulin and hepatocyte growth factor receptors. This protein is involved in endosomal trafficking from the plasma membrane to recycling endosomes or the trans-Golgi network. This gene encodes two transcript variants encoding distinct isoforms.Specifications
DOCK10 | |
Polyclonal | |
Unconjugated | |
DOCK10 | |
dedicator of cytokinesis 10, DOCK10, DRIP2, KIAA0694, Nbla10300, ZIZ3, Zizimin 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
210293, 301556, 55619 | |
-20°C | |
Liquid |
Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BZN6, Q96BY6 | |
DOCK10 | |
DOCK10 Fusion Protein Ag10486 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |