missing translation for 'onlineSavingsMsg'
Läs mer

Dodecanoic acid ingenol ester, MedChemExpress

Produktkod. 16431558
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16431558 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16431558 Leverantör Medchem Express Leverantörsnummer HYN0867

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress Dodecanoic acid ingenol ester is a natural compound.

Dodecanoic acid ingenol ester is a natural compound.

Specifikationer

Kemiskt namn eller material Dodecanoic acid ingenol ester
CAS 54706-70-6
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C32H50O7
Kvantitet 5 mg
LEDER CCCCCCCCCCCC(O[C@@]12C[C@@H](C)[C@@](C=C(C)[C@@H]3O)(C4=O)[C@]3(O)[C@H](O)C(CO)=C[C@@]4([H])[C@]1([H])C2(C)C)=O
Molekylvikt (g/mol) 546.74
Formel vikt 546.74
Procent renhet 99.37%
Kvalitet Research
Fysisk form Solid
Visa mer Visa mindre

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.