Läs mer
Dodecyl Chloroacetate, TRC
Produktkod. 30374676
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30374676 50 mg
1 options
Produktkod. 30374676 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD55246050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dodecyl Chloroacetate
CAS 6316-04-7
Molekylformel C14 H27 Cl O2
Kvantitet 50 mg
Synonym 1-Dodecyl Chloroacetate, Dodecyl 2-Chloroacetate, Lauryl Chloroacetate, NSC 20853, n-Dodecyl Chloroacetate, n-Dodecyl Monochloroacetate
InChI formel InChI=1S/C14H27ClO2/c1-2-3-4-5-6-7-8-9-10-11-12-17-14(16)13-15/h2-13H2,1H3
LEDER CCCCCCCCCCCCOC(=O)CCl
IUPAC-namn dodecyl 2-chloroacetate
Molekylvikt (g/mol) 262.816
Formel vikt 262.17