Läs mer
Dodecyl Hydrogen Maleate, TRC
Produktkod. 30379643
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30379643 500 mg
1 options
Produktkod. 30379643 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD536823500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dodecyl Hydrogen Maleate
CAS 2424-61-5
Molekylformel C16 H28 O4
Kvantitet 500 mg
Synonym 2-Butenedioic acid (Z)-, monododecyl ester, Maleic acid, dodecyl ester (6CI,7CI), Maleic acid, monododecyl ester (8CI), Dodecyl hydrogen maleate, Hydrogen lauryl maleate, Maleic acid monolauryl ester, Monododecyl maleate, Monolauryl maleate
InChI formel InChI=1S/C16H28O4/c1-2-3-4-5-6-7-8-9-10-11-14-20-16(19)13-12-15(17)18/h12-13H,2-11,14H2,1H3,(H,17,18)/b13-12-
LEDER CCCCCCCCCCCCOC(=O)\C=C/C(=O)O
IUPAC-namn (Z)-4-dodecoxy-4-oxobut-2-enoic acid
Molekylvikt (g/mol) 284.391
Formel vikt 284.199