Läs mer
Dodecyl Polyoxyethylene (23) Aldehyde, TRC
Produktkod. 30420534
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30420534 50 mg
30418857 10 mg
30408929 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30420534 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP68942050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 202408-13-7
Molekylformel C14 H28 O2 (C2 H4 O)n
Molekylvikt (g/mol) 272.42
Synonym Brij™ L23 Aldehyde, 2-[2-(Dodecyloxy)ethoxy]acetaldehyde
LEDER CCCCCCCCCCCCOCCOCC=O
Specifikationer
Kemiskt namn eller material Dodecyl Polyoxyethylene (23) Aldehyde
Kvantitet 50 mg
InChI formel InChI=1S/C16H32O3/c1-2-3-4-5-6-7-8-9-10-11-13-18-15-16-19-14-12-17/h12H,2-11,13-16H2,1H3
Formel vikt 272.2351