Läs mer
Dodine, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Dodecylguanidinium Acetate |
| CAS | 2439-10-3 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C13 H29 N3 . C2 H4 O2 |
| Kvantitet | 5 g |
| Synonym | Dodecylguanidinium Acetate, Guanidine, N-dodecyl-, acetate (1:1), Guanidine, dodecyl-, acetate (6CI,7CI), Guanidine, dodecyl-, monoacetate (8CI,9CI), AC 5223, Aadodin, American Cyanamid 5,223, Carpene, Comet, Cyprex, Cyprex 65W, Dodecylguanidine acetate, Dodecylguanidinium acetate, Dodex, Dodex (pesticide), Dodin, Dodine, Dodine 400, Dodine 544SC, Dodine 600SC, Dodine gorsac, Doguadine, Elast, Karpen, Laurylguanidine acetate, Melprex, Melprex 65, Melprex 65W, Melprex Liquid Dodine, Radspor, Syllit, Syllit 65, n-Dodecylguanidine acetate |
| InChI formel | InChI=1S/C13H29N3.C2H4O2/c1-2-3-4-5-6-7-8-9-10-11-12-16-13(14)15;1-2(3)4/h2-12H2,1H3,(H4,14,15,16);1H3,(H,3,4) |
| LEDER | CCCCCCCCCCCCNC(=N)N.CC(=O)O |
| IUPAC-namn | acetic acid;1-dodecylguanidine |
| Molekylvikt (g/mol) | 287.44 |
| Visa mer |