Läs mer
Dofamium Chloride, LoGiCal
Produktkod. 30460337
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30460337 5 mg
1 options
Produktkod. 30460337 Leverantör LoGiCal Leverantörsnummer LGCFOR3067.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dofamium Chloride
CAS 54063-35-3
Analyt- eller komponentnamn Dofamium Chloride
Frakt skick Room Temperature
Molekylformel C25 H44 N3 O2 . Cl
Certifikat och efterlevnad ISO 9001
Synonym Ethanaminium, N,N-dimethyl-N-[2-[methyl(1-oxododecyl)amino]ethyl]-2-oxo-2-(phenylamino)-, chloride (1:1), Desogen (6CI), Dimethyl[2-(N-methyldodecanamido)ethyl][(phenylcarbamoyl)methyl]ammonium chloride (7CI), Ethanaminium, N,N-dimethyl-N-[2-[methyl(1-oxododecyl)amino]ethyl]-2-oxo-2-(phenylamino)-, chloride (9CI), Desocin, Desogen-fest, Dofamium chloride, G 25268
InChI formel InChI=1S/C25H43N3O2.ClH/c1-5-6-7-8-9-10-11-12-16-19-25(30)27(2)20-21-28(3,4)22-24(29)26-23-17-14-13-15-18-23;/h13-15,17-18H,5-12,16,19-22H2,1-4H3;1H
Lösningstyp Neat
LEDER [Cl-].CCCCCCCCCCCC(=O)N(C)CC[N+](C)(C)CC(=O)Nc1ccccc1
IUPAC-namn (2-anilino-2-oxoethyl)-[2-[dodecanoyl(methyl)amino]ethyl]-dimethylazanium;chloride
Industry Type Clinical Testing
Molekylvikt (g/mol) 454.09
Kvantitet 5 mg
Formel vikt 453.3122 g/mol