Läs mer
Dofetilide N-Oxide, TRC
Produktkod. 30374364
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30374364 50 mg
1 options
Produktkod. 30374364 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD52580050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dofetilide N-Oxide
CAS 144449-71-8
Molekylformel C19H27N3O6S2
Kvantitet 50 mg
Synonym N-[4-[2-[Methyl[2-[4-[(methylsulfonyl)amino]phenoxy]ethyl]oxidoamino]ethyl]phenyl]-methanesulfonamide, N-[4-[2-[methyl[2-[4-[(methylsulfonyl)amino]phenoxy]ethyl]amino]ethyl]phenyl]methanesulfonamide N-oxide, UK 116856
InChI formel InChI=1S/C19H27N3O6S2/c1-22(23,13-12-16-4-6-17(7-5-16)20-29(2,24)25)14-15-28-19-10-8-18(9-11-19)21-30(3,26)27/h4-11,20-21H,12-15H2,1-3H3
LEDER O=S(NC1=CC=C(OCCN(CCC2=CC=C(NS(=O)(C)=O)C=C2)(C)=O)C=C1)(C)=O
Molekylvikt (g/mol) 457.56
Formel vikt 457.1341