missing translation for 'onlineSavingsMsg'
Learn More
Learn More
domain-II-of-FIZ1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | domain-II-of-FIZ1 |
---|---|
Concentration | 0.22 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16835194
|
Proteintech
15827-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16825194
|
Proteintech
15827-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
FLJ14768 is homologous to mouse and bovine Fiz1 (flt-3 interacting zinc finger). It interacts with NRL (neural retina leucine zipper). Possible role for Fiz1 as a transcriptional repressor.Specifications
domain-II-of-FIZ1 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q96SL8 | |
84922 | |
Peptide | |
Primary | |
-20°C | |
FIZ1 |
0.22 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
domain I of FIZ 1, domain II of FIZ1, FIZ1, FLT3 interacting zinc finger 1, Zinc finger protein 798, ZNF798 | |
FIZ1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |