Läs mer
(+)-Donepezil, TRC
Produktkod. 30368830
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30368830 10 mg
1 options
Produktkod. 30368830 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD53177010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (+)-Donepezil
CAS 142698-19-9
Molekylformel C24H29NO3
Kvantitet 10 mg
Synonym (R)-2,3-dihydro-5,6-dimethoxy-2-[[1-(phenylmethyl)-4-piperidinyl]methyl]-1H-Inden-1-one, (R)-Donepezil
InChI formel InChI=1S/C24H29NO3/c1-27-22-14-19-13-20(24(26)21(19)15-23(22)28-2)12-17-8-10-25(11-9-17)16-18-6-4-3-5-7-18/h3-7,14-15,17,20H,8-13,16H2,1-2H3/t20-/m1/s1
LEDER O=C1[C@H](CC2CCN(CC3=CC=CC=C3)CC2)CC4=C1C=C(OC)C(OC)=C4
Molekylvikt (g/mol) 379.49