Läs mer
Donitriptan Hydrochloride, TRC
Produktkod. 30007385
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007385 10 mg
30008247 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30007385 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD53250010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 170911-68-9
Molekylformel C23 H25 N5 O2 . H Cl
Molekylvikt (g/mol) 439.94
Synonym 4-[4-[2-[[3-(2-Aminoethyl)-1H-indol-5-yl]oxy]acetyl]-1-piperazinyl]benzonitrile Hydrochloride, 1-[[[3-(2-Aminoethyl)-1H-indol-5-yl]oxy]acetyl]-4-(4-cyanophenyl) piperazine Hydrochloride, F 11356;
IUPAC-namn 4-[4-[2-[[3-(2-aminoethyl)-1H-indol-5-yl]oxy]acetyl]piperazin-1-yl]benzonitrile;hydrochloride
LEDER Cl.NCCc1c[nH]c2ccc(OCC(=O)N3CCN(CC3)c4ccc(cc4)C#N)cc12
Specifikationer
Kemiskt namn eller material Donitriptan Hydrochloride
Kvantitet 10 mg
InChI formel InChI=1S/C23H25N5O2.ClH/c24-8-7-18-15-26-22-6-5-20(13-21(18)22)30-16-23(29)28-11-9-27(10-12-28)19-3-1-17(14-25)2-4-19;/h1-6,13,15,26H,7-12,16,24H2;1H
Formel vikt 439.1775