missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Donkey anti-Sheep IgG (H+L) Secondary Antibody, Texas Red (Pre-adsorbed), Novus Biologicals™
Donkey Polyclonal Antibody
Brand: Novus Biologicals NBP1-73688
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Sheep samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Specifikationer
| IgG (H+L) | |
| Polyclonal | |
| Texas Red | |
| Donkey | |
| Affinity Purified | |
| RUO | |
| Secondary | |
| Reconstitute with 1.0 ml deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| LYOPH | |
| Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
| Sheep IgG whole molecule | |
| 1 mg | |
| Adaptive Immunity, Immunology | |
| This antibody was pre-adsorbed against Chicken, Guinea Pig, Hamster, Horse, Human, Mouse, Rabbit, and Rat Serum Proteins. No reaction was observed against Chicken, Guinea Pig, Hamster, Horse, Human, Mouse, Rabbit and Rat Serum Proteins. | |
| Sheep | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering