missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Dopamine Transporter Monoclonal Antibody (6V-23-23)

Beskrivning
This antibody does not cross-react with human samples.
The Dopamine Transporter (DAT) is responsible for the reaccumulation of dopamine after it has been released. DAT antibodies and antibodies for other markers of catecholamine biosynthesis are widely used as markers for dopaminergic and noradrenergic neurons in a variety of applications including depression, schizophrenia, Parkinson's disease and drug abuse.
Specifikationer
Specifikationer
| Antigen | Dopamine Transporter |
| Användningsområden | Western Blot, Immunocytochemistry |
| Klassificering | Monoclonal |
| Klona | 6V-23-23 |
| Koncentration | 1.62 mg/mL |
| Konjugera | Unconjugated |
| Formulering | 0.02M potassium phosphate with 0.15M NaCl and 0.1% sodium azide; pH 7.2 |
| Gen | Slc6a3 |
| Genaccessionsnr. | P23977, Q01959, Q61327 |
| Gene Alias | DA transporter; Dat; Dat1; dopamine transporter; dopamine transporter 1; PKDYS; SLC6A3; Sodium-dependent dopamine transporter; solute carrier family 6 (neurotransmitter transporter), member 3; solute carrier family 6 (neurotransmitter transporter, dopamine), member 3; solute carrier family 6 member 3; solute carrier family 6, member 3 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?