Läs mer
Dorzolamide N-Sulfonamide Dimer, TRC
Produktkod. 30368829
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30368829 5 mg
1 options
Produktkod. 30368829 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD5351105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dorzolamide N-Sulfonamide Dimer
CAS 199734-72-0
Molekylformel C20H29N3O8S6
Kvantitet 5 mg
Synonym (4R,6R)-rel-4-(Ethylamino)-N-[[(4S,6S)-4-(ethylamino)-5,6-dihydro-6-methyl-7,7-dioxido-4H-thieno[2,3-b]thiopyran-2-yl]sulfonyl]-5,6-dihydro-6-methyl-4H-thieno[2,3-b]thiopyran-2-sulfonamide-7,7-dioxide
InChI formel InChI=1S/C20H29N3O8S6/c1-5-21-15-7-11(3)34(24,25)19-13(15)9-17(32-19)36(28,29)23-37(30,31)18-10-14-16(22-6-2)8-12(4)35(26,27)20(14)33-18/h9-12,15-16,21-23H,5-8H2,1-4H3/t11-,12-,15-,16-/m0/s1
LEDER CCN[C@H]1C[C@H](C)S(=O)(=O)c2sc(cc12)S(=O)(=O)NS(=O)(=O)c3cc4[C@H](C[C@H](C)S(=O)(=O)c4s3)NCC
Molekylvikt (g/mol) 631.85