Läs mer
DOTA-NHS-ester Hexafluorophosphate TFA Salt (Technical Grade), TRC
Produktkod. 30006706
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006706 50 mg
30010528 10 mg
30010835 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30006706 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD49486750MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1823122-52-6
Molekylformel C20H31N5O10 . C2HF3O2 . HPF6
Synonym 1,4,7,10-Tetraazacyclododecane-N,N',N' ',N' ' '-tetraacetic acid mono(N-hydroxysuccinimidyl) ester Hexafluorophosphate TFA Salt, 1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid mono(N-hydroxysuccinimide ester) Hexafluorophosphate TFA Salt
LEDER OC(=O)CN1CCN(CC(=O)O)CCN(CC(=O)ON2C(=O)CCC2=O)CCN(CC(=O)O)CC1.OC(=O)C(F)(F)F.F[PH](F)(F)(F)(F)F
Specifikationer
Kemiskt namn eller material DOTA-NHS-ester Hexafluorophosphate TFA Salt (Technical Grade)
Kvantitet 50 mg
InChI formel InChI=1S/C20H31N5O10.C2HF3O2.F6HP/c26-15-1-2-16(27)25(15)35-20(34)14-24-9-7-22(12-18(30)31)5-3-21(11-17(28)29)4-6-23(8-10-24)13-19(32)33;3-2(4,5)1(6)7;1-7(2,3,4,5)6/h1-14H2,(H,28,29)(H,30,31)(H,32,33);(H,6,7);7H
Formel vikt 761.172