Läs mer
Dotinurad, TRC
Produktkod. 30364208
Change view
Kvantitet:
0.5 mg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30364208 1 mg
30365163 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30364208 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2626751MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1285572-51-1
Molekylformel C14H9Cl2NO4S
Molekylvikt (g/mol) 358.19
LEDER Oc1c(Cl)cc(cc1Cl)C(=O)N2CS(=O)(=O)c3ccccc23
Specifikationer
Kemiskt namn eller material Dotinurad
Kvantitet 1 mg
InChI formel InChI=1S/C14H9Cl2NO4S/c15-9-5-8(6-10(16)13(9)18)14(19)17-7-22(20,21)12-4-2-1-3-11(12)17/h1-6,18H,7H2
Formel vikt 356.9629