Läs mer
DOTP, TRC
Produktkod. 30383009
Change view
Kvantitet:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30383009 250 mg
30385700 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30383009 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD494890250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 91987-74-5
Molekylformel C12H32N4O12P4
Molekylvikt (g/mol) 548.3
Synonym 1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetra(methylene Phosphonic Acid), 1,4,7,10-Tetraazacyclododecane, Phosphonic Acid deriv., 1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetramethylenephosphonic Acid, DOTMP, NSC 681106, N,N',N'',N'''-Tetrakis(phosphonomethyl)-1,4,7,10-tetraazacyclododecane
LEDER OP(=O)(O)CN1CCN(CP(=O)(O)O)CCN(CP(=O)(O)O)CCN(CP(=O)(O)O)CC1
Specifikationer
Kemiskt namn eller material DOTP
Kvantitet 250 mg
InChI formel InChI=1S/C12H32N4O12P4/c17-29(18,19)9-13-1-2-14(10-30(20,21)22)5-6-16(12-32(26,27)28)8-7-15(4-3-13)11-31(23,24)25/h1-12H2,(H2,17,18,19)(H2,20,21,22)(H2,23,24,25)(H2,26,27,28)