Learn More
Metro™ Double-Deep qwikTRAK™ Mobile Unit Kits
Click to view available options
Width (English):
18 in.
21 in.
24 in.
Width (English) Shelf:
18 in.
21 in.
24 in.
Finish:
Chrome
Heavy Polymer
Metroseal Green Epoxy
Stainless Steel
Unit Size:
Each
Description
- For use with Metro Super Erecta™, Super Erecta Pro™ Super Adjustable™ and MetroMax i™ shelving systems
- Kit includes eight 74 in. (188cm) high -UP posts, four V-groove casters; one stainless steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware
Order Info
Order one kit per double-deep system. Shelves are sold separately.
Specifications
Specifications
Product Type | Mobile Unit Kit |
Finish | Metroseal Green Epoxy |
For Use With (Equipment) | Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ shelving system |
Includes | Eight 74 in. high up posts, four V-groove casters, one Stainless Steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware |
Width (English) Shelf | 24 in. |
Width (English) | 24 in. |
Width (Metric) Shelf | 61 cm |
Width (Metric) | 61 cm |
Material | Steel |
Product Content Correction
Product Title
Metro™ Double-Deep qwikTRAK™ Mobile Unit Kits >
Spot an opportunity for improvement?Share a Content Correction