Learn More
BRAND™ Double-Ended PTFE Magnetic Stirring Bars
Excellent centering
Brand:  BRAND™ 137930
2191.16 SEK valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 0.11160kg
 View more versions of this product
Product Code. 12025292
  • 2665.00 SEK / Pack of 10
Estimated Shipment: 08-05-2024
to see stock.
Add to basket
Description
Description
  • Small contact surface
  • High turbulence
  • Works well even at low speeds
  • Specifications
    Specifications
    8 mm
    10
    37 mm
    Dumbbell