Läs mer
VELP Scientifica™ Double Rod Mini H-Stand Set
Produktkod. 17110206 Handla allt Velp Produkter
Change view
Mått (L x B x H):
300 x 430 mm
Produktkod. Mått (L x B x H)
17110206 300 x 430 mm
1 options
Produktkod. 17110206 Leverantör VELP Scientifica™ Leverantörsnummer A00000383
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
The double rod mini H-stand set for disperser tools is designed for light weight instruments and comes complete. This stand is designed to avoid rotation of attachments and instruments.
DESCRIPTION
  • Designed for light weight instruments and comes complete with all parts
  • Designed to avoid rotation of attachment and instrument through the use of the double rod and bosshead clamp
Specifikationer
Produkttyp Stand
Mått (L x B x H) 300 x 430 mm
Autoklaveringsbar Not Autoclavable