Läs mer
Dowex Marathon C, H+ form, Thermo Scientific™
Kvantitet:
250 g
1000 g
2500 g
Förpackningsstorlek:
1000g
2500g
250g
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is designed for demineralization using HCl or H2 SO4 as regenerate in co-current or counter-current operation.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
It is designed for demineralization using HCl or H2 SO4 as regenerate in co-current or counter-current operation.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Dowex Marathon C |
| Namnnotering | H+ form |
| CAS | 150604-77-6 |
| MDL-nummer | MFCD00212649 |
| Densitet | 1.2 |
| Kvantitet | 2500 g |
| Löslighetsinformation | Insoluble in water. |
Korrigering av produktinnehåll
Produkttitel
Hittar du en möjlighet till förbättring?Übermitteln Sie eine inhaltliche Korrektur