Läs mer
Doxepin EP Impurity A, Mikromol™
Produktkod. 30462996
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30462996 4 x 25 mg
17924346 100 mg
17961916 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30462996 Leverantör Mikromol Leverantörsnummer MM0032.0300254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Doxepin EP Impurity A, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dibenzo[b,e]oxepin-11(6H)-one (Doxepinone)
CAS 4504-87-4
Analyt- eller komponentnamn Dibenzo[b,e]oxepin-11(6H)-one (Doxepinone)
Frakt skick Room Temperature
Molekylformel C14 H10 O2
Certifikat och efterlevnad ISO 9001
Synonym Doxepin Hydrochloride Imp. A (EP), Doxepin Imp. A (EP), Doxepin USP Related Compound A, Doxepin USP RC A, Dibenzo[b,e]oxepin-11(6H)-one, Doxepinone, Doxepin Related Compound A, Doxepin Hydrochloride Impurity A, Doxepin Impurity A
InChI formel InChI=1S/C14H10O2/c15-14-11-6-2-1-5-10(11)9-16-13-8-4-3-7-12(13)14/h1-8H,9H2
Lösningstyp Neat
LEDER O=C1c2ccccc2COc3ccccc13
IUPAC-namn 6H-benzo[c][1]benzoxepin-11-one
Molekylvikt (g/mol) 210.23
Kvantitet 4 x 25 mg
Formel vikt 210.0681 g/mol