Läs mer
Doxorubicin-hydrazone-caproyl-maleimide, TRC
Produktkod. 30369544
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30369544 5 mg
1 options
Produktkod. 30369544 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD5580405MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Doxorubicin-hydrazone-caproyl-maleimide
CAS 151038-96-9
Molekylformel C37H42N4O13
Kvantitet 5 mg
Synonym DOXO-EMCH, Doxorubicin-EMCH, (E)-N'-(1-((2S,4S)-4-(((2R,4S,5S,6S)-4-Amino-5-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-2,5,12-trihydroxy-7-methoxy-6,11-dioxo-1,2,3,4,6,11-hexahydrotetracen-2-yl)-2-hydroxyethylidene)-6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanehydrazide
InChI formel InChI=1S/C37H42N4O13/c1-17-32(46)20(38)13-27(53-17)54-22-15-37(51,23(16-42)39-40-24(43)9-4-3-5-12-41-25(44)10-11-26(41)45)14-19-29(22)36(50)31-30(34(19)48)33(47)18-7-6-8-21(52-2)28(18)35(31)49/h6-8,10-11,17,20,22,27,32,42,46,48,50-51H,3-5,9,12-16,38H2,1-2H3,(H,40,43)/t17-,20-,22-,27-,32+,37-/m0/s1
LEDER OC=1C2=C([C@@H](O[C@H]3C[C@H](N)[C@H](O)[C@H](C)O3)C[C@@](C(=NNC(CCCCCN4C(=O)C=CC4=O)=O)CO)(O)C2)C(O)=C5C1C(=O)C=6C(C5=O)=C(OC)C=CC6
IUPAC-namn N'-(1-((2S,4S)-4-(((2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-2,5,12-trihydroxy-7-methoxy-6,11-dioxo-1,2,3,4,6,11-hexahydrotetracen-2-yl)-2-hydroxyethylidene)-6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanehydrazide
Molekylvikt (g/mol) 750.75
Formel vikt 750.27