Läs mer
Doxybetasol, TRC
Produktkod. 30379760
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30379760 10 mg
30383952 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30379760 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD56100010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1879-77-2
Molekylformel C22 H29 F O4
Molekylvikt (g/mol) 376.46
Synonym (11β,16β)-9-Fluoro-11,17-dihydroxy-16-methyl-pregna-1,4-diene-3,20-dione, Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17-dihydroxy-16β-methyl- (7CI,8CI), 21-Deoxybetamethasone, 9-Fluoro-11β,17-dihydroxy-16β-methylpregna-1,4-diene-3,20-dione, Doxibetasol, Doxybetasol, Betamethasone Valerate Imp. B (EP)
IUPAC-namn (8S,9R,10S,11S,13S,14S,16S,17R)-17-acetyl-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
LEDER C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)C
Specifikationer
Kemiskt namn eller material Doxybetasol
Kvantitet 10 mg
InChI formel InChI=1S/C22H29FO4/c1-12-9-17-16-6-5-14-10-15(25)7-8-19(14,3)21(16,23)18(26)11-20(17,4)22(12,27)13(2)24/h7-8,10,12,16-18,26-27H,5-6,9,11H2,1-4H3/t12-,16-,17-,18-,19-,20-,21-,22-/m0/s1
Formel vikt 376.205