Läs mer
Doxycycline 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifikationer
Specifikationer
| Kemiskt namn eller material | Doxycycline |
| CAS | 564-25-0 |
| Analyt- eller komponentnamn | Doxycycline |
| Frakt skick | +20°C |
| Molekylformel | C22 H24 N2 O9 |
| Koncentration | 1000 μg/mL |
| Certifikat och efterlevnad | ISO 17034 |
| Synonym | 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, (4S,4aR,5S,5aR,6R,12aS)-, 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo- (6CI,8CI), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, [4S-(4α,4aα,5α,5aα,6α,12aα)]-, (4S,4aR,5S,5aR,6R,12aS)-4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide, (-)-Doxycycline, 4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide, 5-Hydroxy-α-6-deoxytetracycline, 6-Deoxy-5-hydroxytetracycline, 6-Deoxyoxytetracycline, Bidox-DT, Ceedox, Dentistar, Deoxymykoin, Dotur, Doxicip, Doximal, Doxinyl, Doxivetin, Doxy RW, Doxy-Gel, Doxycen, Doxycycline, Doxymycin, Doxytetracycline, Emdox, GS 3065, Hydramycin, Idoxy, Lenteclin, Liviatin, Lupidox, Medeomycin, Monodox, Oxytetracycline, 6-deoxy-, Pdox-LB, Polodoksin, Pulmodox, R-Doxy, Ronaxan, Tetradox, Tolexine, Unidox, Vibramycin, Vibramycine, Vibranos SF, Vibraveineuse, Vibravenos, α-6-Deoxy-5-hydroxytetracycline, α-6-Deoxyoxytetracycline, α-Doxycycline |
| InChI formel | InChI=1S/C22H24N2O8/c1-7-8-5-4-6-9(25)11(8)16(26)12-10(7)17(27)14-15(24(2)3)18(28)13(21(23)31)20(30)22(14,32)19(12)29/h4-7,10,14-15,17,25,27-29,32H,1-3H3,(H2,23,31)/t7-,10+,14+,15-,17-,22-/m0/s1 |
| LEDER | C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H](N(C)C)C(=C(C(=O)N)C(=O)[C@@]3(O)C(=C2C(=O)c4c(O)cccc14)O)O |
| Visa mer |