missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doxycycline hyclate, USP grade, MP Biomedicals™
Click to view available options
Quantity:
1 g
5 g
Specifications
Type | Doxycycline Hyclate |
Physical Form | Powder |
Formula Weight | 1025.89 |
Grade | USP |
Color | Yellow |
Quantity | 5 g |
Chemical Name or Material | Doxycycline hyclate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'