missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Doxycycline monohydrate, 97%

Catalog Number p-7077763
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
17086-28-1
C22H24N2O8·H2O
462.45
FZKWRPSUNUOXKJ-CVHRZJFOSA-N
doxycycline monohydrate, doxycycline hydrate, adoxa, doxycycline hcl, 4-dimethylamino-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrate, adoxa pak, 4s,4ar,5s,5ar,6r,12as-4-dimethylamino-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrate, vibramycin-d, doxycycline usp, monodox tn
54684461
(4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrate
CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O
This item is not returnable. View return policy
17086-28-1
C22H24N2O8·H2O
462.45
FZKWRPSUNUOXKJ-CVHRZJFOSA-N
doxycycline monohydrate, doxycycline hydrate, adoxa, doxycycline hcl, 4-dimethylamino-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrate, adoxa pak, 4s,4ar,5s,5ar,6r,12as-4-dimethylamino-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrate, vibramycin-d, doxycycline usp, monodox tn
54684461
(4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrate
CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 167.0°C to 168.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 462.45
Percent Purity 97%
Chemical Name or Material Doxycycline monohydrate
compliance-icons
Product Identifier
  • Doxycycline monohydrate
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361d-Suspected of damaging the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'