Learn More
Doxycycline monohydrate, 97%, ACROS Organics™
Brand:  Acros Organics 458890250
Additional Details : CAS Number : 17086-28-1 Weight : 0.02500kg
Quantity 25g
 View more versions of this product
Product Code. 15580594
  • 2578.00 SEK / 25g
Estimated Shipment
14-12-2020
Specifications
Chemical Identifiers
17086-28-1
462.45
doxycycline monohydrate, doxycycline hydrate, adoxa, doxycycline hcl, 4-dimethylamino-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrate, adoxa pak, 4s,4ar,5s,5ar,6r,12as-4-dimethylamino-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrate, vibramycin-d, doxycycline usp, monodox tn
(4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrate
C22H24N2O8·H2O
FZKWRPSUNUOXKJ-CVHRZJFOSA-N
54684461
CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O
Specifications
Doxycycline monohydrate
3.5 to 4.5%
25g
96% min. (HPLC)
doxycycline monohydrate, doxycycline hydrate, adoxa, doxycycline hcl, 4-dimethylamino-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrate, adoxa pak, 4s,4ar,5s,5ar,6r,12as-4-dimethylamino-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrate, vibramycin-d, doxycycline usp, monodox tn
CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O
462.45
462.45
Authentic
167.0°C to 168.0°C
17086-28-1
C22H24N2O8·H2O
FZKWRPSUNUOXKJ-CVHRZJFOSA-N
(4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrate
54684461
97%
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Doxycycline monohydrate
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361d-Suspected of damaging the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.