Läs mer
Doxylamine Beta-D-Glucuronide (Mixture of diastereomers), TRC
Produktkod. 30371085
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30371085 10 mg
30370550 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30371085 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD56201010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 137908-79-3
Molekylformel C23H30N2O7
Molekylvikt (g/mol) 446.5
Synonym N-beta-D-Glucopyranuronosyl-N,N-dimethyl-2-[1-phenyl-1-(2-pyridinyl)ethoxy]ethanaminium Inner Salt
LEDER CC(OCC[N+](C)(C)C1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(=O)[O-])(c2ccccc2)c3ccccn3
Specifikationer
Kemiskt namn eller material Doxylamine beta-D-Glucuronide (Mixture of diastereomers)
Kvantitet 10 mg
InChI formel InChI=1S/C23H30N2O7/c1-23(15-9-5-4-6-10-15,16-11-7-8-12-24-16)31-14-13-25(2,3)21-19(28)17(26)18(27)20(32-21)22(29)30/h4-12,17-21,26-28H,13-14H2,1-3H3/t17-,18-,19+,20-,21?,23?/m0/s1
Formel vikt 446.2053