Läs mer
Doxylamine N'-Oxide, TRC
Produktkod. 17943845
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17943845 100 mg
17935115 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17943845 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD562030100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 99430-77-0
Molekylformel C17H22N2O2
Molekylvikt (g/mol) 286.37
Synonym N,N-Dimethyl-2-[1-(1-oxido-2-pyridinyl)-1-phenylethoxy]-ethanamine, N,N-Dimethyl-2-[1-phenyl-1-(2-pyridinyl)ethoxy]-ethanamine N2-Oxide
LEDER O=N1=C(C(OCCN(C)C)(C)C2=CC=CC=C2)C=CC=C1
Specifikationer
Kemiskt namn eller material Doxylamine N'-Oxide
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C17H22N2O2/c1-17(21-14-13-18(2)3,15-9-5-4-6-10-15)16-11-7-8-12-19(16)20/h4-12H,13-14H2,1-3H3
Formel vikt 286.1681
Procent renhet >95%