Läs mer
Lovibond™ DPD #3 tablets
Produktkod. 11958031
Kvantitet:
500 tabletter
Produktkod. 11958031
Brand: Lovibond™ 511082BT
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
The suitable tablet for the fast result
Water analysis couldn't be easier. Precise dosing, long shelf life and easy handling make the tablet reagent the most popular indicator system and the cheapest way to have analyse a parameter.
The tablet reagent in the aluminum blister pack withstands almost all climatic conditions and external influences. In addition, there is no safety risk for the user.
Specifikationer
Kvantitet 500 tabletter
Räckvidd DPD No. 3
Typ Reagens