missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | DPF2 |
---|---|
Concentration | 0.16 mg/mL |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16839633
|
Proteintech
12111-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16829633
|
Proteintech
12111-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the d4 domain family, characterized by a zinc finger-like structural motif. This protein functions as a transcription factor which is necessary for the apoptotic response following deprivation of survival factors. It likely serves a regulatory role in rapid hematopoietic cell growth and turnover. This gene is considered a candidate gene for multiple endocrine neoplasia type I, an inherited cancer syndrome involving multiple parathyroid, enteropancreatic, and pituitary tumors.Specifications
DPF2 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q61103, Q92785 | |
19708, 361711, 5977 | |
DPF2 Fusion Protein Ag2756 | |
Primary | |
-20°C | |
DPF2 |
0.16 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
BAF45D, BRG1 associated factor 45D, DPF2, Protein requiem, REQ, ubi d4, UBID4, Zinc finger protein ubi d4 | |
Dpf2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |