Läs mer
DPN, MedChemExpress
Beskrivning
DPN (Diarylpropionitrile) is a non-steroidal estrogen receptor β (ERβ) selective ligand, with an EC50 of 0.85 nM. DPN has neuroprotective effects in a number of neurological diseases.
Specifikationer
Specifikationer
| Kemiskt namn eller material | DPN |
| CAS | 1428-67-7 |
| Färg | White |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C15H13NO2 |
| Kvantitet | 10 mg |
| Synonym | Diarylpropionitrile |
| Löslighetsinformation | DMSO : 100 mg/mL (417.94 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (2.80 mM; Need ultrasonic) |
| LEDER | OC1=CC=C(CC(C#N)C2=CC=C(O)C=C2)C=C1 |
| Molekylvikt (g/mol) | 239.27 |
| Visa mer |